Assay Details
Gene Symbol: LOC5572212 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001653830.2 | XP_001653880.2 | 6 - 7 | 6195 | 60 | |
XM_021843085.1 | XP_021698777.1 | 7 - 8 | 6007 | 60 | ||
XM_021843086.1 | XP_021698778.1 | 7 - 8 | 5961 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() |
