Assay Details
Gene Symbol: LOC5571159 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021854228.1 | XP_021709920.1 | 8 - 9 | 2067 | 74 | |
XM_021854229.1 | XP_021709921.1 | 8 - 9 | 2054 | 74 | ||
XM_021854230.1 | XP_021709922.1 | 8 - 9 | 2042 | 74 | ||
XM_021854231.1 | XP_021709923.1 | 8 - 9 | 1969 | 74 | ||
XM_021854232.1 | XP_021709924.1 | 8 - 9 | 2029 | 74 | ||
XM_021854233.1 | XP_021709925.1 | 8 - 9 | 1963 | 74 | ||
XM_021854234.1 | XP_021709926.1 | 6 - 7 | 1672 | 74 | ||
XM_021854235.1 | XP_021709927.1 | 8 - 9 | 1965 | 74 | ||
XM_021854236.1 | XP_021709928.1 | 7 - 8 | 1802 | 74 | ||
XM_021854237.1 | XP_021709929.1 | 8 - 9 | 1954 | 74 | ||
XM_021854238.1 | XP_021709930.1 | 8 - 9 | 1866 | 74 | ||
XM_021854239.1 | XP_021709931.1 | 8 - 9 | 2520 | 74 | ||
XM_021854240.1 | XP_021709932.1 | 8 - 9 | 2005 | 74 | ||
XM_021854241.1 | XP_021709933.1 | 8 - 9 | 2510 | 74 | ||
XM_021854242.1 | XP_021709934.1 | 8 - 9 | 2453 | 74 | ||
XM_021854243.1 | XP_021709935.1 | 7 - 8 | 1870 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() |
