Assay Details
Gene Symbol: LOC5571159 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021854228.1 | XP_021709920.1 | 6 - 7 | 1789 | 83 | |
XM_021854229.1 | XP_021709921.1 | 6 - 7 | 1776 | 83 | ||
XM_021854230.1 | XP_021709922.1 | 6 - 7 | 1764 | 83 | ||
XM_021854231.1 | XP_021709923.1 | 6 - 7 | 1691 | 83 | ||
XM_021854232.1 | XP_021709924.1 | 6 - 7 | 1751 | 83 | ||
XM_021854233.1 | XP_021709925.1 | 6 - 7 | 1685 | 83 | ||
XM_021854234.1 | XP_021709926.1 | 4 - 5 | 1394 | 83 | ||
XM_021854235.1 | XP_021709927.1 | 6 - 7 | 1687 | 83 | ||
XM_021854236.1 | XP_021709928.1 | 5 - 6 | 1524 | 83 | ||
XM_021854237.1 | XP_021709929.1 | 6 - 7 | 1676 | 83 | ||
XM_021854238.1 | XP_021709930.1 | 6 - 7 | 1588 | 83 | ||
XM_021854239.1 | XP_021709931.1 | 6 - 7 | 2242 | 83 | ||
XM_021854240.1 | XP_021709932.1 | 6 - 7 | 1727 | 83 | ||
XM_021854241.1 | XP_021709933.1 | 6 - 7 | 2232 | 83 | ||
XM_021854242.1 | XP_021709934.1 | 6 - 7 | 2175 | 83 | ||
XM_021854243.1 | XP_021709935.1 | 5 - 6 | 1592 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() |
