Assay Details
Gene Symbol: LOC5571159 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021854228.1 | XP_021709920.1 | 18 - 19 | 4341 | 70 | |
XM_021854229.1 | XP_021709921.1 | 18 - 19 | 4328 | 70 | ||
XM_021854230.1 | XP_021709922.1 | 18 - 19 | 4316 | 70 | ||
XM_021854231.1 | XP_021709923.1 | 18 - 19 | 4243 | 70 | ||
XM_021854232.1 | XP_021709924.1 | 18 - 19 | 4303 | 70 | ||
XM_021854233.1 | XP_021709925.1 | 18 - 19 | 4237 | 70 | ||
XM_021854234.1 | XP_021709926.1 | 16 - 17 | 3946 | 70 | ||
XM_021854235.1 | XP_021709927.1 | 18 - 19 | 4239 | 70 | ||
XM_021854236.1 | XP_021709928.1 | 17 - 18 | 4076 | 70 | ||
XM_021854237.1 | XP_021709929.1 | 18 - 19 | 4228 | 70 | ||
XM_021854238.1 | XP_021709930.1 | 18 - 19 | 4140 | 70 | ||
XM_021854239.1 | XP_021709931.1 | 18 - 19 | 4794 | 70 | ||
XM_021854240.1 | XP_021709932.1 | 18 - 19 | 4279 | 70 | ||
XM_021854241.1 | XP_021709933.1 | 18 - 19 | 4784 | 70 | ||
XM_021854242.1 | XP_021709934.1 | 18 - 19 | 4727 | 70 | ||
XM_021854243.1 | XP_021709935.1 | 17 - 18 | 4144 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
