Assay Details
Gene Symbol: LOC5571159 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021854228.1 | XP_021709920.1 | 11 - 12 | 2901 | 108 | |
XM_021854229.1 | XP_021709921.1 | 11 - 12 | 2888 | 108 | ||
XM_021854230.1 | XP_021709922.1 | 11 - 12 | 2876 | 108 | ||
XM_021854231.1 | XP_021709923.1 | 11 - 12 | 2803 | 108 | ||
XM_021854232.1 | XP_021709924.1 | 11 - 12 | 2863 | 108 | ||
XM_021854233.1 | XP_021709925.1 | 11 - 12 | 2797 | 108 | ||
XM_021854234.1 | XP_021709926.1 | 9 - 10 | 2506 | 108 | ||
XM_021854235.1 | XP_021709927.1 | 11 - 12 | 2799 | 108 | ||
XM_021854236.1 | XP_021709928.1 | 10 - 11 | 2636 | 108 | ||
XM_021854237.1 | XP_021709929.1 | 11 - 12 | 2788 | 108 | ||
XM_021854238.1 | XP_021709930.1 | 11 - 12 | 2700 | 108 | ||
XM_021854239.1 | XP_021709931.1 | 11 - 12 | 3354 | 108 | ||
XM_021854240.1 | XP_021709932.1 | 11 - 12 | 2839 | 108 | ||
XM_021854241.1 | XP_021709933.1 | 11 - 12 | 3344 | 108 | ||
XM_021854242.1 | XP_021709934.1 | 11 - 12 | 3287 | 108 | ||
XM_021854243.1 | XP_021709935.1 | 10 - 11 | 2704 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() |
