Assay Details
Gene Symbol: LOC5564524 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021848242.1 | XP_021703934.1 | 10 - 11 | 5324 | 76 | |
XM_021848243.1 | XP_021703935.1 | 10 - 11 | 5459 | 76 | ||
XM_021848244.1 | XP_021703936.1 | 10 - 11 | 5369 | 76 | ||
XM_021848245.1 | XP_021703937.1 | 10 - 11 | 5246 | 76 | ||
XM_021848246.1 | XP_021703938.1 | 10 - 11 | 5278 | 76 | ||
XM_021848247.1 | XP_021703939.1 | 10 - 11 | 5309 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() |
