Assay Details
Gene Symbol: LOC5567482 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021852554.1 | XP_021708246.1 | 2 - 3 | 773 | 66 | |
XM_021852555.1 | XP_021708247.1 | 2 - 3 | 585 | 66 | ||
XM_021852556.1 | XP_021708248.1 | 2 - 3 | 538 | 66 | ||
XM_021852557.1 | XP_021708249.1 | 2 - 3 | 599 | 66 | ||
XM_021852558.1 | XP_021708250.1 | 2 - 3 | 383 | 66 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021852554.1, XM_021852555.1, XM_021852556.1, XM_021852557.1, XM_021852558.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
