Assay Details
Gene Symbol: LOC5578157 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021844714.1 | XP_021700406.1 | 5 - 6 | 2026 | 78 | |
XM_021844717.1 | XP_021700409.1 | 5 - 6 | 1620 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
