Assay Details
Gene Symbol: LOC5567649 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001651793.2 | XP_001651843.2 | 6 - 7 | 1402 | 90 | |
XM_021844638.1 | XP_021700330.1 | 7 - 8 | 1848 | 90 | ||
XM_021844639.1 | XP_021700331.1 | 6 - 7 | 1817 | 90 | ||
XM_021844641.1 | XP_021700333.1 | 7 - 8 | 1847 | 90 | ||
XM_021844642.1 | XP_021700334.1 | 6 - 7 | 1402 | 90 |
