Assay Details
Gene Symbol: LOC5570905 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001653333.2 | XP_001653383.2 | 11 - 12 | 2603 | 84 | |
XM_021844949.1 | XP_021700641.1 | 12 - 13 | 2680 | 84 | ||
XM_021844951.1 | XP_021700643.1 | 11 - 12 | 2532 | 84 | ||
XM_021844952.1 | XP_021700644.1 | 12 - 13 | 2700 | 84 | ||
XM_021844953.1 | XP_021700645.1 | 12 - 13 | 2699 | 84 | ||
XM_021844954.1 | XP_021700646.1 | 11 - 12 | 2514 | 84 | ||
XM_021844955.1 | XP_021700647.1 | 11 - 12 | 2326 | 84 | ||
XM_021844956.1 | XP_021700648.1 | 11 - 12 | 2112 | 84 | ||
XM_021844957.1 | XP_021700649.1 | 11 - 12 | 2047 | 84 | ||
XM_021844958.1 | XP_021700650.1 | 11 - 12 | 2120 | 84 | ||
XM_021844959.1 | XP_021700651.1 | 11 - 12 | 2060 | 84 |
