Assay Details
Gene Symbol: LOC5570905 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001653333.2 | XP_001653383.2 | 6 - 7 | 1770 | 58 | |
XM_021844949.1 | XP_021700641.1 | 7 - 8 | 1847 | 58 | ||
XM_021844951.1 | XP_021700643.1 | 6 - 7 | 1699 | 58 | ||
XM_021844952.1 | XP_021700644.1 | 7 - 8 | 1867 | 58 | ||
XM_021844953.1 | XP_021700645.1 | 7 - 8 | 1866 | 58 | ||
XM_021844954.1 | XP_021700646.1 | 6 - 7 | 1681 | 58 | ||
XM_021844955.1 | XP_021700647.1 | 6 - 7 | 1493 | 58 | ||
XM_021844956.1 | XP_021700648.1 | 6 - 7 | 1279 | 58 | ||
XM_021844957.1 | XP_021700649.1 | 6 - 7 | 1214 | 58 | ||
XM_021844958.1 | XP_021700650.1 | 6 - 7 | 1287 | 58 | ||
XM_021844959.1 | XP_021700651.1 | 6 - 7 | 1227 | 58 |

More Information
Set Membership: |
![]() ![]() |
