Assay Details
Gene Symbol: LOC5571582 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021851752.1 | XP_021707444.1 | 3 - 4 | 2411 | 66 | |
XM_021851753.1 | XP_021707445.1 | 3 - 4 | 2843 | 66 |

More Information
Set Membership: |
![]() ![]() |
