Assay Details
Gene Symbol: LOC5571582 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001659741.2 | XP_001659791.1 | 4 - 5 | 2667 | 101 | |
XM_021851752.1 | XP_021707444.1 | 4 - 5 | 2682 | 101 | ||
XM_021851753.1 | XP_021707445.1 | 4 - 5 | 3114 | 101 |

More Information
Set Membership: |
![]() ![]() |
