Assay Details
Gene Symbol: LOC5574815 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001655325.2 | XP_001655375.2 | 1 - 2 | 188 | 80 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001655325.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
