Assay Details
Gene Symbol: LOC5569376 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001658400.2 | XP_001658450.1 | 2 - 3 | 252 | 110 | |
XM_021844770.1 | XP_021700462.1 | 4 - 5 | 813 | 110 | ||
XM_021844771.1 | XP_021700463.1 | 4 - 5 | 869 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() |
