Assay Details
Gene Symbol: LOC5569376 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021844770.1 | XP_021700462.1 | 3 - 4 | 616 | 105 | |
XM_021844771.1 | XP_021700463.1 | 3 - 4 | 672 | 105 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021844770.1, XM_021844771.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
