Assay Details
Gene Symbol: LOC5566218 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001650571.2 | XP_001650621.2 | 2 - 3 | 1125 | 68 | |
XM_021838781.1 | XP_021694473.1 | 2 - 3 | 695 | 68 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021838781.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
