Assay Details
Gene Symbol: LOC5577645 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021838140.1 | XP_021693832.1 | 2 - 3 | 884 | 90 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021838140.1 transcripts.
Set Membership: |
![]() ![]() |
