Assay Details
Gene Symbol: LOC5577645 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021838137.1 | XP_021693829.1 | 6 - 7 | 2890 | 56 | |
XM_021838138.1 | XP_021693830.1 | 6 - 7 | 2986 | 56 | ||
XM_021838139.1 | XP_021693831.1 | 6 - 7 | 2768 | 56 | ||
XM_021838140.1 | XP_021693832.1 | 6 - 7 | 2863 | 56 | ||
XM_021838141.1 | XP_021693833.1 | 6 - 7 | 2800 | 56 | ||
XM_021838142.1 | XP_021693834.1 | 6 - 7 | 2773 | 56 | ||
XM_021838143.1 | XP_021693835.1 | 6 - 7 | 2315 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() |
