Assay Details
Gene Symbol: LOC5577645 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021838137.1 | XP_021693829.1 | 7 - 8 | 2962 | 72 | |
XM_021838138.1 | XP_021693830.1 | 7 - 8 | 3058 | 72 | ||
XM_021838139.1 | XP_021693831.1 | 7 - 8 | 2840 | 72 | ||
XM_021838140.1 | XP_021693832.1 | 7 - 8 | 2935 | 72 | ||
XM_021838141.1 | XP_021693833.1 | 7 - 8 | 2872 | 72 | ||
XM_021838142.1 | XP_021693834.1 | 7 - 8 | 2845 | 72 | ||
XM_021838143.1 | XP_021693835.1 | 7 - 8 | 2387 | 72 |
