Assay Details
Gene Symbol: LOC5578803 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021854000.1 | XP_021709692.1 | 2 - 3 | 2606 | 61 | |
XM_021854002.1 | XP_021709694.1 | 3 - 4 | 1765 | 61 | ||
XM_021854003.1 | XP_021709695.1 | 3 - 4 | 2058 | 61 | ||
XM_021854004.1 | XP_021709696.1 | 3 - 4 | 1634 | 61 | ||
XM_021854005.1 | XP_021709697.1 | 2 - 3 | 1346 | 61 | ||
GenBank mRNA | L46373.1 | - | 1 - 2 | 776 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
