Assay Details
Gene Symbol: LOC5578803 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021854000.1 | XP_021709692.1 | 11 - 12 | 4389 | 70 | |
XM_021854002.1 | XP_021709694.1 | 12 - 13 | 3548 | 70 | ||
XM_021854003.1 | XP_021709695.1 | 12 - 13 | 3841 | 70 | ||
XM_021854004.1 | XP_021709696.1 | 12 - 13 | 3417 | 70 | ||
XM_021854005.1 | XP_021709697.1 | 11 - 12 | 3129 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
