Assay Details
Gene Symbol: LOC5578803 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021854000.1 | XP_021709692.1 | 10 - 11 | 4294 | 72 | |
XM_021854002.1 | XP_021709694.1 | 11 - 12 | 3453 | 72 | ||
XM_021854003.1 | XP_021709695.1 | 11 - 12 | 3746 | 72 | ||
XM_021854004.1 | XP_021709696.1 | 11 - 12 | 3322 | 72 | ||
XM_021854005.1 | XP_021709697.1 | 10 - 11 | 3034 | 72 | ||
GenBank mRNA | L46373.1 | - | 9 - 10 | 2464 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() |
