Assay Details
Gene Symbol: LOC5578803 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021854000.1 | XP_021709692.1 | 5 - 6 | 3432 | 62 | |
XM_021854002.1 | XP_021709694.1 | 6 - 7 | 2591 | 62 | ||
XM_021854003.1 | XP_021709695.1 | 6 - 7 | 2884 | 62 | ||
XM_021854004.1 | XP_021709696.1 | 6 - 7 | 2460 | 62 | ||
XM_021854005.1 | XP_021709697.1 | 5 - 6 | 2172 | 62 | ||
GenBank mRNA | L46373.1 | - | 4 - 5 | 1602 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() |
