Assay Details
Gene Symbol: LOC5578803 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021854000.1 | XP_021709692.1 | 3 - 4 | 2828 | 81 | |
XM_021854002.1 | XP_021709694.1 | 4 - 5 | 1987 | 81 | ||
XM_021854003.1 | XP_021709695.1 | 4 - 5 | 2280 | 81 | ||
XM_021854004.1 | XP_021709696.1 | 4 - 5 | 1856 | 81 | ||
XM_021854005.1 | XP_021709697.1 | 3 - 4 | 1568 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() |
