Assay Details
Gene Symbol: LOC5578537 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001663878.2 | XP_001663928.2 | 5 - 6 | 1374 | 79 | |
XM_021841579.1 | XP_021697271.1 | 5 - 6 | 1397 | 79 | ||
XM_021841580.1 | XP_021697272.1 | 6 - 7 | 1331 | 79 | ||
XM_021841581.1 | XP_021697273.1 | 5 - 6 | 835 | 79 |
