Assay Details
Gene Symbol: LOC5578537 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001663878.2 | XP_001663928.2 | 4 - 5 | 1301 | 71 | |
XM_021841579.1 | XP_021697271.1 | 4 - 5 | 1324 | 71 | ||
XM_021841580.1 | XP_021697272.1 | 5 - 6 | 1258 | 71 | ||
XM_021841581.1 | XP_021697273.1 | 4 - 5 | 762 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() |
