Assay Details
Gene Symbol: LOC5578537 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001663878.2 | XP_001663928.2 | 1 - 2 | 718 | 69 | |
XM_021841579.1 | XP_021697271.1 | 1 - 2 | 720 | 69 | ||
XM_021841580.1 | XP_021697272.1 | 1 - 2 | 720 | 69 |

More Information
Set Membership: |
![]() ![]() |
