Assay Details
Gene Symbol: LOC5571502 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001659702.2 | XP_001659752.1 | 4 - 5 | 526 | 105 | |
XM_021848426.1 | XP_021704118.1 | 4 - 5 | 593 | 105 | ||
XM_021848427.1 | XP_021704119.1 | 4 - 5 | 604 | 105 | ||
XM_021848434.1 | XP_021704126.1 | 4 - 5 | 526 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
