Assay Details
Gene Symbol: LOC5578342 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001656861.2 | XP_001656911.2 | 2 - 3 | 2400 | 112 | |
XM_021849011.1 | XP_021704703.1 | 3 - 4 | 2648 | 112 | ||
XM_021849012.1 | XP_021704704.1 | 3 - 4 | 2604 | 112 | ||
XM_021849013.1 | XP_021704705.1 | 3 - 4 | 2592 | 112 |

More Information
Set Membership: |
![]() ![]() |
