Assay Details
Gene Symbol: LOC5563793 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021846334.1 | XP_021702026.1 | 5 - 6 | 5267 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC5563793 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021846334.1 | XP_021702026.1 | 5 - 6 | 5267 | 63 |
Set Membership: |
![]() ![]() ![]() ![]() |