Assay Details
Gene Symbol: LOC5571510 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021848408.1 | XP_021704100.1 | 10 - 11 | 2400 | 81 | |
XM_021848409.1 | XP_021704101.1 | 10 - 11 | 2382 | 81 | ||
XM_021848410.1 | XP_021704102.1 | 9 - 10 | 2269 | 81 | ||
XM_021848411.1 | XP_021704103.1 | 10 - 11 | 3365 | 81 | ||
XM_021848412.1 | XP_021704104.1 | 10 - 11 | 2057 | 81 | ||
XM_021848414.1 | XP_021704106.1 | 10 - 11 | 1766 | 81 | ||
XM_021848415.1 | XP_021704107.1 | 10 - 11 | 2050 | 81 | ||
XM_021848416.1 | XP_021704108.1 | 10 - 11 | 2042 | 81 | ||
XM_021848417.1 | XP_021704109.1 | 10 - 11 | 1798 | 81 |
