Assay Details
Gene Symbol: LOC5571510 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021848408.1 | XP_021704100.1 | 11 - 12 | 2557 | 81 | |
XM_021848409.1 | XP_021704101.1 | 11 - 12 | 2539 | 81 | ||
XM_021848410.1 | XP_021704102.1 | 10 - 11 | 2426 | 81 | ||
XM_021848411.1 | XP_021704103.1 | 11 - 12 | 3522 | 81 | ||
XM_021848412.1 | XP_021704104.1 | 11 - 12 | 2214 | 81 | ||
XM_021848414.1 | XP_021704106.1 | 11 - 12 | 1923 | 81 | ||
XM_021848415.1 | XP_021704107.1 | 11 - 12 | 2207 | 81 | ||
XM_021848416.1 | XP_021704108.1 | 11 - 12 | 2199 | 81 | ||
XM_021848417.1 | XP_021704109.1 | 11 - 12 | 1955 | 81 |
