Assay Details
Gene Symbol: LOC5571510 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021848414.1 | XP_021704106.1 | 2 - 3 | 456 | 87 | |
XM_021848415.1 | XP_021704107.1 | 2 - 3 | 740 | 87 | ||
XM_021848416.1 | XP_021704108.1 | 2 - 3 | 732 | 87 | ||
XM_021848417.1 | XP_021704109.1 | 2 - 3 | 488 | 87 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021848414.1, XM_021848415.1, XM_021848416.1, XM_021848417.1 transcripts.
Set Membership: |
![]() ![]() |
