Assay Details
Gene Symbol: LOC5571510 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021848408.1 | XP_021704100.1 | 3 - 4 | 1274 | 79 | |
XM_021848410.1 | XP_021704102.1 | 2 - 3 | 1143 | 79 | ||
XM_021848411.1 | XP_021704103.1 | 3 - 4 | 2239 | 79 | ||
XM_021848412.1 | XP_021704104.1 | 3 - 4 | 931 | 79 | ||
XM_021848414.1 | XP_021704106.1 | 3 - 4 | 640 | 79 | ||
XM_021848415.1 | XP_021704107.1 | 3 - 4 | 924 | 79 | ||
XM_021848416.1 | XP_021704108.1 | 3 - 4 | 916 | 79 | ||
XM_021848417.1 | XP_021704109.1 | 3 - 4 | 672 | 79 |
