Assay Details
Gene Symbol: LOC5571510 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021848408.1 | XP_021704100.1 | 9 - 10 | 2269 | 110 | |
XM_021848409.1 | XP_021704101.1 | 9 - 10 | 2251 | 110 | ||
XM_021848410.1 | XP_021704102.1 | 8 - 9 | 2138 | 110 | ||
XM_021848411.1 | XP_021704103.1 | 9 - 10 | 3234 | 110 | ||
XM_021848412.1 | XP_021704104.1 | 9 - 10 | 1926 | 110 | ||
XM_021848414.1 | XP_021704106.1 | 9 - 10 | 1635 | 110 | ||
XM_021848415.1 | XP_021704107.1 | 9 - 10 | 1919 | 110 | ||
XM_021848416.1 | XP_021704108.1 | 9 - 10 | 1911 | 110 | ||
XM_021848417.1 | XP_021704109.1 | 9 - 10 | 1667 | 110 |

More Information
Set Membership: |
![]() ![]() |
