Assay Details
Gene Symbol: LOC5577066 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021840649.1 | XP_021696341.1 | 1 - 2 | 45 | 84 | |
XM_021840652.1 | XP_021696344.1 | 1 - 2 | 45 | 84 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021840649.1, XM_021840652.1 transcripts.
Set Membership: |
![]() ![]() |
