Assay Details
Gene Symbol: LOC5577066 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021840649.1 | XP_021696341.1 | 2 - 3 | 390 | 74 | |
XM_021840650.1 | XP_021696342.1 | 2 - 3 | 663 | 74 | ||
XM_021840651.1 | XP_021696343.1 | 2 - 3 | 477 | 74 | ||
XM_021840653.1 | XP_021696345.1 | 2 - 3 | 522 | 74 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021840649.1, XM_021840650.1, XM_021840651.1, XM_021840653.1 transcripts.
Set Membership: |
![]() ![]() |
