Assay Details
Gene Symbol: LOC5571294 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021840150.1 | XP_021695842.1 | 6 - 7 | 5054 | 62 | |
XM_021840153.1 | XP_021695845.1 | 6 - 7 | 5054 | 62 |

More Information
Set Membership: |
![]() ![]() |
