Assay Details
Gene Symbol: LOC5571294 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021840149.1 | XP_021695841.1 | 13 - 14 | 6421 | 59 | |
XM_021840150.1 | XP_021695842.1 | 12 - 13 | 6373 | 59 | ||
XM_021840152.1 | XP_021695844.1 | 12 - 13 | 6358 | 59 | ||
XM_021840153.1 | XP_021695845.1 | 11 - 12 | 6310 | 59 | ||
XM_021840154.1 | XP_021695846.1 | 13 - 14 | 6214 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() |
