Assay Details
Gene Symbol: LOC5571294 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021840149.1 | XP_021695841.1 | 11 - 12 | 5816 | 68 | |
XM_021840150.1 | XP_021695842.1 | 10 - 11 | 5768 | 68 | ||
XM_021840152.1 | XP_021695844.1 | 10 - 11 | 5753 | 68 | ||
XM_021840153.1 | XP_021695845.1 | 9 - 10 | 5705 | 68 |

More Information
Set Membership: |
![]() ![]() |
