Assay Details
Gene Symbol: LOC5571294 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021840152.1 | XP_021695844.1 | 9 - 10 | 5281 | 70 | |
XM_021840153.1 | XP_021695845.1 | 8 - 9 | 5233 | 70 |

More Information
Set Membership: |
![]() ![]() |
