Assay Details
Gene Symbol: LOC5573316 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001654486.2 | XP_001654536.2 | 2 - 3 | 617 | 62 | |
XM_021837751.1 | XP_021693443.1 | 2 - 3 | 496 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() |
