Assay Details
Gene Symbol: LOC5573316 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001654486.2 | XP_001654536.2 | 3 - 4 | 2841 | 94 | |
XM_021837751.1 | XP_021693443.1 | 3 - 4 | 2720 | 94 |
