Assay Details
Gene Symbol: LOC5571904 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021855131.1 | XP_021710823.1 | 3 - 4 | 844 | 61 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021855131.1 transcripts.
Set Membership: |
![]() ![]() |
