Assay Details
Gene Symbol: LOC5578528 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001663869.2 | XP_001663919.2 | 7 - 8 | 2013 | 70 | |
XM_021855093.1 | XP_021710785.1 | 7 - 8 | 2046 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
