Assay Details
Gene Symbol: LOC5574073 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001661096.2 | XP_001661146.1 | 1 - 2 | 514 | 92 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001661096.2 transcripts.
Set Membership: |
![]() ![]() |
