Assay Details
Gene Symbol: LOC5575136 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001655463.2 | XP_001655513.1 | 7 - 8 | 1370 | 61 | |
XM_021842214.1 | XP_021697906.1 | 7 - 8 | 1370 | 61 | ||
XM_021842215.1 | XP_021697907.1 | 6 - 7 | 1211 | 61 | ||
XM_021842216.1 | XP_021697908.1 | 6 - 7 | 1043 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() |
