Assay Details
Gene Symbol: LOC5575136 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001655463.2 | XP_001655513.1 | 9 - 10 | 1917 | 68 | |
XM_021842214.1 | XP_021697906.1 | 9 - 10 | 1902 | 68 | ||
XM_021842215.1 | XP_021697907.1 | 8 - 9 | 1758 | 68 | ||
XM_021842216.1 | XP_021697908.1 | 8 - 9 | 1590 | 68 | ||
XM_021842217.1 | XP_021697909.1 | 8 - 9 | 1524 | 68 | ||
XM_021842218.1 | XP_021697910.1 | 8 - 9 | 1509 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
