Assay Details
Gene Symbol: LOC5571585 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021849062.1 | XP_021704754.1 | 2 - 3 | 394 | 96 | |
XM_021849063.1 | XP_021704755.1 | 2 - 3 | 433 | 96 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021849062.1, XM_021849063.1 transcripts.
Set Membership: |
![]() ![]() |
